ContactUsLinkComponent
Accessories for Signature™ Ultra-Low Temperature Upright Freezers, –86 to –50°C, Rack Packages with 2 Shelves of Sliding Drawer Racks and Boxes
Accessories for Signature™ Ultra-Low Temperature Upright Freezers, –86 to –50°C, Rack Packages with 2 Shelves of Sliding Drawer Racks and Boxes
  • Catalog No:
  • CA89200-182
  • CA89200-180
  • 89423-568
  • CA89200-176
  • CA89200-174
  • CA89200-172
  • CA89200-170
  • CA89200-168
  • CA89200-210
  • CA89200-208
  • CA89200-206
  • CA89200-110
  • 25000-136
  • 89423-534
  • 25000-134
  • 89423-542
  • 89423-536
  • 89423-538
  • CA89200-204
  • 89423-544
  • CA89200-202
  • 89423-546
  • CA89200-188
  • CA89200-118
  • CA89200-114
  • CA89200-128
  • 89423-554
  • 89423-552
  • 89423-562
  • 89423-558
  • 89423-560
  • Cat. No.:
  • CA89200-182
  • CA89200-180
  • 89423-568
  • CA89200-176
  • CA89200-174
  • CA89200-172
  • CA89200-170
  • CA89200-168
  • CA89200-210
  • CA89200-208
  • CA89200-206
  • CA89200-110
  • 25000-136
  • 89423-534
  • 25000-134
  • 89423-542
  • 89423-536
  • 89423-538
  • CA89200-204
  • 89423-544
  • CA89200-202
  • 89423-546
  • CA89200-188
  • CA89200-118
  • CA89200-114
  • CA89200-128
  • 89423-554
  • 89423-552
  • 89423-562
  • 89423-558
  • 89423-560
  • Specifications:
  • Upright Rack with Adjustable Shelves for 490L (17.3 cu. ft.) and 651L (23 cu. ft.) Freezers
  • Upright Rack with Adjustable Shelves for 490L (17.3 cu. ft.) and 651L (23 cu. ft.) Freezers
  • Holds 42 Deep Well Microplates; fits 793L (28 cu. ft.), 4 Inner Door Freezers; 5.3W x 11.96H x 25.8"D
  • Upright Rack for 793L (28 cu. ft.) Freezer
  • Upright Rack with Adjustable Shelves for 368L (13 cu. ft.) Freezer
  • Upright Rack with Adjustable Shelves for 368L (13 cu. ft.) Freezer
  • Upright Rack for 490L (17.3 cu. ft.) and 651L (23 cu. ft.) Freezers
  • Upright Rack for 368L (13 cu. ft.) Freezer
  • Contains 1 Shelf of Sliding Drawer Racks (4 Racks)
  • Holds 32 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 1.082"H; fits 368L (13 cu. ft.), 4 Inner Door Freezers
  • Holds 24 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 1.082"H; fits 368L (13 cu. ft.), 4 Inner Door Freezers
  • Holds 48 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 1.082"H; fits 490L (17.3 cu. ft.) and 651L (23 cu. ft.), 4 Inner Door Freezers
  • Holds 56 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 1.082"H; fits 793L (28 cu. ft.), 4 Inner Door Freezers
  • Holds 36 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 1.082"H; fits 490L (17.3 cu. ft.) and 651L (23 cu. ft.), 4 Inner Door Freezers
  • Holds 42 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 1.082"H; fits 793L (28 cu. ft.), 4 Inner Door Freezers
  • Upright Rack for 490L (17.3 cu. ft.) and 651L (23 cu. ft.) Freezers
  • Contains 1 Shelf of Sliding Drawer Racks (5 Racks)
  • Contains 1 Shelf of Sliding Drawer Racks (4 Racks)
  • Contains 1 Shelf of Sliding Drawer Racks (6 Racks)
  • Holds 28 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 2.3"H; fits 793L (28 cu. ft.), 4 Inner Door Freezers
  • Holds 24 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 2.3"H; fits 490L (17.3 cu. ft.) and 651L (23 cu. ft.), 4 Inner Door Freezers
  • Holds 21 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 3.78"H; fits 793L (28 cu. ft.), 4 Inner Door Freezers
  • Holds 12 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 3.78"H; fits 368L (13 cu. ft.), 4 Inner Door Freezers
  • Holds 18 Thermo Scientific Tube Boxes; 3.365W x 5.03L x 3.78"H; fits 490L (17.3 cu. ft.) and 651L (23 cu. ft.), 4 Inner Door Freezers
Frequently Bought Together