ContactUsLinkComponent
Ward's® AP Chemistry Investigation 7: Stoichiometry: What is the Greenest Way to Separate a Mixture?
  470303-038
undefined
Ward's® AP Chemistry Investigation 7: Stoichiometry: What is the Greenest Way to Separate a Mixture?
  470303-038
 :  470303-038
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Ward's® AP Chemistry Investigation 7: Stoichiometry
  • No. of Lab Groups:
    15
  • Cat. no.:
    470303-038
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier No.:
    470303-038
Meets AP Science Practices 2, 4, 5, and 6
  • Addresses Big Idea 3
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, students will gain deeper understanding of "green" chemistry, stoichiometry, and the peer review process. Students will investigate how to purify a mixture using green chemistry and then work through the peer review process to determine which submitted manuscript is the most complete and would be appropriate for publication in a hypothetical scientific journal. Includes contaminated mixture. Requires Bunsen burner, striker, iron ring, ceramic triangle, ring stand/clamp, crucible/lid, balance, and crucible tongs, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
 : Activity includes Analysis Mixture. Materials required but not provided are Bunsen Burner/tubing, Striker, Fuel, Iron Ring, Ceramic Triangles, Ring Stands, Crucibles/Lids, Crucible tongs, Collection container, Analytical Digital Balances, Transferring scoops, Lab Notebook, Gloves, Safety Goggles, Lab Aprons, and Hot Pads.