ContactUsLinkComponent
Ward's® AP Chemistry Investigation 3: Gravimetric Analysis: How Hard is the Water?
Catalog # 470303-030
Supplier:  Ward's Science
undefined
Ward's® AP Chemistry Investigation 3: Gravimetric Analysis: How Hard is the Water?
Catalog # 470303-030
Supplier:  Ward's Science
Supplier Number:  470303-030
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ward's® AP Chemistry Investigation 3: Gravimetric Analysis
  • No. of Lab Groups:
    15
  • Cat. no.:
    470303-030
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier No.:
    470303-030
Specifications
About this item
Meets AP Science Practices 1, 2, 4, 5, 6, and 7
  • Addresses Big Idea 1, 2, & 3
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
The central challenge in this kit is determining the hardness of a water sample. Students will be tasked with designing a procedure in the guided inquiry section to determine the hardness of water. Students are tasked with determining which of six water samples, representing different areas in their community, require a water softening system. Gravimetric Analysis is the central technique used to analyze the sample. Includes Calcium chloride and sodium carbonate. Requires a 250 mL Buchner funnel, No. 6 Buchner funnel rubber stopper, aspirator or vacuum pump, rubber tubing, analytical balance, drying oven, and ring stand/clamps, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Ordering information: Kit contents include Calcium Chloride Anhydrous, Sodium Carbonate Anhydrous, Sodium Carbonate, and Calcium Chloride. Materials required but not provided are DI Water, Beakers, Graduated Cylinders, Watch Glasses, Erlenmeyer Flasks, Rubber Stoppers, Buchner Funnels, Side-Arm Flasks, Vacuum pump or aspirator, Filter paper, Funnel, 3 inch glass tubing (optional), Ring Stands, Clamps, Analytical Balances, Drying Oven (optional), Rubber Tubing, Pipettes, Stir Rods, Lab Notebook, Gloves, Safety Goggles, and Lab Aprons.