ContactUsLinkComponent
Ward's® AP Chemistry Investigation 1: Spectroscopy: What is the Concentration of that Solution?
Ward's® AP Chemistry Investigation 1: Spectroscopy: What is the Concentration of that Solution?
Catalog # 470303-012
Supplier:  Ward's Science
undefined
Ward's® AP Chemistry Investigation 1: Spectroscopy: What is the Concentration of that Solution?
Catalog # 470303-012
Supplier:  Ward's Science
Supplier Number:  470303-012
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ward's® AP Chemistry Investigation 1: Spectroscopy
  • No. of Lab Groups:
    15
  • Cat. no.:
    470303-012
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier No.:
    470303-012
Specifications
About this item
Meets AP Science Practices 2, 4, 5, and 6
  • Addresses Big Idea 1
  • Material provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this kit students will determine the concentration of blue dye in an unknown solution called Scienceade. A calibration line will be made with a known sample and this will be used as an aid in solving the problem. Students will need to justify the selection of a particular type of spectroscopy to measure properties associated with vibrational or electronic motions of molecules and they will design and interpret the results of an experiment regarding the absorption of light to determine the concentration of an absorbing species in a solution. Includes blue dye no.1 and Scienceade Stock Solution. Requires spectrophotometer or colorimeter, and cuvettes, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Ordering information: Kit contents include Blue Dye No. 1, Scienceade, and instructions. Materials required but not provided are Glass or plastic cuvettes, DI Water, Graduated cylinders, Pipettes, Spectrophotometer or Colorimeter, Test tubes, Tape or marker, Gloves, Safety Goggles, Aprons, and Graphing Software.