Order Entry
Canada
ContactUsLinkComponent
Calpain Inhibitor III
Calpain Inhibitor III
Supplier:  Bachem Americas
Certificates
Calpain Inhibitor III
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    88191-84-8
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MDLMFCD00798796
  • Protein/peptide name:
    Calpain Inhibitor III
  • Molecular weight:
    382.46
  • Sequence:
    Z-Val-Phe-aldehyde
  • Molecular formula:
    C₂₂H₂₆N₂O₄

Specifications

Frequently Bought Together