Mca-Pro-Leu-Gly-Leu-Glu-Glu-Ala-Dap(Dnp)-NH₂ Trifluoroacetate
Highly selective MMP-12 FRET substrate with a kcat/Km value of 1.85 · 10⁵ M⁻¹s⁻¹. Poor substrate of other MMPs with the exception of MMP-13 (kcat/Km = 0.53 · 10 ⁵ M⁻¹s⁻¹) and MMP-9 (0.33 · 10⁵ M⁻¹s⁻¹).
- CAS No.:891198-38-2
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782572
- Protein/peptide name:Mca-Pro-Leu-Gly-Leu-Glu-Glu-Ala-Dap(Dnp)-NH₂ Trifluoroacetate
- Molecular weight:1195.21
- Sequence:Mca-Pro-Leu-Gly-Leu-Glu-Glu-Ala-Dap(Dnp)-NH₂
- Molecular formula:C₅₃H₇₀N₁₂O₂₀
Frequently Bought Together