(Z-Asp-Glu-Val-Asp)₂-Rhodamine 110
Fournisseur: Bachem Americas
Certificats
À propos de cet article
(Z-DEVD)₂-R110, highly sensitive, photostable fluorogenic substrate for the determination of intracellular caspase-3 activity. The fluorophore Rhodamine 110 (R110) can be detected at an emission wavelength of 535 nm using an excitation wavelength of 485 nm. As with (H-Asp-Glu-Val-Asp)₂-Rhodamine 110 (M-2620) the fluorescence intensity can be measured by flow cytometry, microwell plate reader, or fluorescence microscopy.
Spécifications
- CAS No.:223538-61-2
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD08702756
- Protein/peptide name:(Z-Asp-Glu-Val-Asp)₂-Rhodamine 110
- Molecular weight:1515.46
- Sequence:(Z-Asp-Glu-Val-Asp)₂-Rhodamine 110
- Molecular formula:C₇₂H₇₈N₁₀O₂₇