Abz-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (669-674)-EDDnp Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
Intramolecularly quenched fluorescent substrate containing the ortho-aminobenzoyl (Abz) / N-(2,4-dinitrophenyl)ethylenediamine (EDDnp) groups as the donor / acceptor pair. It corresponds to the Swedish-mutated (JMV2236) β-amyloid precursor protein (βAPP) sequence targeted by β-secretase BACE (β-site APP-cleaving activity). This FRET substrate is more selectively cleaved by BACE1 and BACE2 than by cathepsin D, a disintegrin and metalloprotease 10 (ADAM10), tumor necrosis α-converting enzyme (TACE), presenilin-1 (PS1), or presenilin-2 (PS2).
Specifications
- CAS No.:1007306-20-8
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD06411972
- Protein/peptide name:Abz-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (669-674)-EDDnp Trifluoroacetate
- Molecular weight:986.99
- Sequence:Abz-Val-Asn-Leu-Asp-Ala-Glu-EDDnp
- Molecular formula:C₄₂H₅₈N₁₂O₁₆