Mca-Amyloid beta/A4 Protein Precursor₇₇₀ (667-676)-Lys(Dnp)-Arg-Arg amide Trifluoroacetate
This fluorescent (FRET) peptide substrate contains the wild-type amyloid precursor protein (APP) β-secretase cleavage site. Mca-SEVKMDAEFRK(Dnp)RR- amide has been used for assaying β-secretase-like activity of thimet oligopeptidase (TOP, EC 3.4.24.15). The results suggested that TOP is a potential β-secretase candidate and is involved in the processing of APP in vivo. See also L-1905.
- CAS No.:1802078-33-6
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782559
- Protein/peptide name:Mca-Amyloid beta/A4 Protein Precursor₇₇₀ (667-676)-Lys(Dnp)-Arg-Arg amide Trifluoroacetate
- Molecular weight:2033.21
- Sequence:Mca-Ser-Glu-Val-Lys-Met-Asp-Ala-Glu-Phe-Arg-Lys(Dnp)-Arg-Arg-NH₂
- Molecular formula:C₈₇H₁₂₉N₂₇O₂₈S
Frequently Bought Together