Mca-(Ala⁷,Lys(Dnp)⁹)-Bradykinin Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
Substrate V, Mca-RPPGFSAFK(Dnp), is a very sensitive internally quenched fluorescent substrate for endothelin-converting enzyme-1 (ECE-1) a membrane-bound zinc metallopeptidase that is related to neprilysin in amino acid sequence. The kcat/Km value for the hydrolysis by ECE-1 was 1.9 x 10⁷ M⁻¹s⁻¹ and 1.7 x 10⁶ M⁻¹s⁻¹ for the hydrolysis by neprilysin. This FRET substrate was hydrolyzed rapidly by stromelysin 1 (MMP-3) with kcat/Km= 218000 M⁻¹s⁻¹ and very slowly by gelatinase B (MMP-9) (kcat/Km= 10100 M⁻¹s⁻¹). There was no hydrolysis by MMP-1 and MMP-2. Mca-RPPGFSAFK(Dnp) has also found use as substrate for insulin-degrading enzyme (IDE) and ACE2.
Specifications
- CAS No.:323577-36-2
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782552
- Protein/peptide name:Mca-(Ala⁷,Lys(Dnp)⁹)-Bradykinin Trifluoroacetate
- Molecular weight:1388.46
- Sequence:Mca-Arg-Pro-Pro-Gly-Phe-Ser-Ala-Phe-Lys(Dnp)-OH
- Molecular formula:C₆₆H₈₁N₁₅O₁₉