Mca-Pro-Leu-Gly-Leu-Dap(Dnp)-Ala-Arg-NH₂ Trifluoroacetate
Fournisseur: Bachem Americas
Certificats
À propos de cet article
Mca-PLGL-Dpa-AR-amide is a very sensitive FRET substrate for continuous assays and in situ determination of the matrix metalloproteinase activity. Cleavage at the Gly-Leu bond separates the highly fluorescent Mca group from the efficient 2,4-dinitrophenyl quencher, resulting in an increase in fluorescence intensity. The kcat/Km values for the punctuated metalloproteinase (MMP-7) and for gelatinase (MMP-2), e.g., are 1.7·10⁵ and 6.3·10⁵ M⁻¹s⁻¹, respectively.
Spécifications
- CAS No.:140430-53-1
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD03844448
- Protein/peptide name:Mca-Pro-Leu-Gly-Leu-Dap(Dnp)-Ala-Arg-NH₂ Trifluoroacetate
- Molecular weight:1093.16
- Sequence:Mca-Pro-Leu-Gly-Leu-Dap(Dnp)-Ala-Arg-NH₂
- Molecular formula:C₄₉H₆₈N₁₄O₁₅