Pz-Pro-Leu-Gly-Pro-D-Arg-OH Trifluoroacetate
See also M-2460 and M-2465.
Chromogenic substrate for the assay of collagenase and thimet oligopeptidase (Pz-peptidase, metalloendopeptidase 24.15, collagenase-like peptidase). Enzymatic cleavage yields Pz-Pro-Leu-OH, G-2970 (or Pz-Pro-OH, E-2275). The cleavage can be monitored by analytical HPLC, detection at 320 nm.
- CAS No.:17011-78-8
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782532
- Protein/peptide name:Pz-Pro-Leu-Gly-Pro-D-Arg-OH Trifluoroacetate
- Molecular weight:776.89
- Sequence:Pz-Pro-Leu-Gly-Pro-D-Arg-OH
- Molecular formula:C₃₈H₅₂N₁₀O₈
Frequently Bought Together