(Trp⁴)-Kemptide
Chocktide ((Trp⁴)-Kemptide) is readily phosphorylated by cAMP-dependent protein kinase. The phosphorylated peptide, in turn, is an effective substrate for phosphoprotein phosphatase. The changes in the substrate fluorescence intensity as a function of its phosphorylation state provide a sensitive assay of both enzyme activities.
- CAS No.:80224-16-4
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00133448
- Protein/peptide name:(Trp⁴)-Kemptide
- Molecular weight:887.05
- Sequence:H-Leu-Arg-Arg-Trp-Ser-Leu-Gly-OH
- Molecular formula:C₄₀H₆₆N₁₄O₉
Frequently Bought Together