H-Gly-Pro-pNA · HCl
Fournisseur: Bachem Americas
Certificats
À propos de cet article
See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .
Excellent substrate for dipeptidylpeptidase IV (DPP IV).
Spécifications
- CAS No.:103213-34-9
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00058170
- Protein/peptide name:H-Gly-Pro-pNA · HCl
- Molecular weight:328.76
- Sequence:H-Gly-Pro-pNA · HCl
- Molecular formula:C₁₃H₁₆N₄O₄ · HCl