Suc-Ala-Ala-Val-pNA
L-1405.0050BA
:
- CAS No.:61043-47-8
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038473
- Protein/peptide name:Suc-Ala-Ala-Val-pNA
- Molecular weight:479.49
- Sequence:Suc-Ala-Ala-Val-pNA
- Molecular formula:C₂₁H₂₉N₅O₈
- Cat. no.:L-1405.0050BA
- Supplier No.:L-1405.0050
See also C-1040, C-2345, I-1000, and L-1435.
Suc-AAV-pNA (SAAVNA), a very specific substrate for human and rat leukocyte elastases.