Z-Ala-Pro-pNA
Catalogue # L-1215.0250BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:66382-56-7
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00238411
- Protein/peptide name:Z-Ala-Pro-pNA
- Molecular weight:440.46
- Sequence:Z-Ala-Pro-pNA
- Molecular formula:C₂₂H₂₄N₄O₆
- Cat. no.:L-1215.0250BA
- Supplier No.:L-1215.0250
Spécifications
À propos de cet article
See also I-1350 and K-1490.
Z-AP-pNA was used in combination with Z-GP-pNA (L-1325) and Z-AA-pNA (L-1495) for characterizing the prolyl oligopeptide of of the hyperthermophile Pyrococcus furiosus.