H-Gly-Asp-Asp-Asp-Asp-Lys-betaNA
Fournisseur: Bachem Americas
Certificats
À propos de cet article
GDDDDK-βNA, specific substrate for the determination of enteropeptidase (enterokinase), also used in histochemistry.
Spécifications
- CAS No.:70023-02-8
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD06798938
- Protein/peptide name:H-Gly-Asp-Asp-Asp-Asp-Lys-betaNA
- Molecular weight:788.77
- Sequence:H-Gly-Asp-Asp-Asp-Asp-Lys-βNA
- Molecular formula:C₃₄H₄₄N₈O₁₄