H-Glu-Glu-betaNA
K-1250.0250BA
:
- CAS No.:23645-15-0
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237877
- Protein/peptide name:H-Glu-Glu-betaNA
- Molecular weight:401.42
- Sequence:H-Glu-Glu-βNA
- Molecular formula:C₂₀H₂₃N₃O₆
- Cat. no.:K-1250.0250BA
- Supplier No.:K-1250.0250