ContactUsLinkComponent
Ac-Arg-Leu-Arg-AMC Trifluoroacetate
Ac-Arg-Leu-Arg-AMC Trifluoroacetate
  • CAS No.:
    929903-87-7
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782518
  • Protein/peptide name:
    Ac-Arg-Leu-Arg-AMC Trifluoroacetate
  • Molecular weight:
    642.76
  • Sequence:
    Ac-Arg-Leu-Arg-AMC
  • Molecular formula:
    C₃₀H₄₆N₁₀O₆
Frequently Bought Together