Boc-Gly-Gly-Gly-OH
Fournisseur: Bachem Americas
Certificats
À propos de cet article
For Boc-dipeptides Boc-Xaa-Gly-OH and Boc-Xaa-Pro-OH, which can be used as building blocks during Boc-SPPS, please see the product family 'Boc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. This family also includes short protected peptide fragments suitable for solution synthesis and preparation of enzyme substrates and inhibitors.
Boc-triglycine.
Spécifications
- CAS No.:28320-73-2
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00190687
- Protein/peptide name:Boc-Gly-Gly-Gly-OH
- Molecular weight:289.29
- Sequence:Boc-Gly-Gly-Gly-OH
- Molecular formula:C₁₁H₁₉N₃O₆