Ammonia Ionic Strength Adjustor, 500 mL, Hach
Ammonia Ionic Strength Adjustor, 500 mL, Hach
Ammonia Ionic Strength Adjustor (ISA), 500 ml, Plastic bottle
Catalog # CA11017-066
Supplier:   Hach
Supplier #:  2824349
Ammonia Ionic Strength Adjustor, 500 mL, Hach
Catalog # CA11017-066
Supplier:  Hach
CAS Number:  
Ammonia Ionic Strength Adjustor, 500 mL, Hach
Ammonia Ionic Strength Adjustor, 500 mL, Hach
Ammonia Ionic Strength Adjustor (ISA), 500 ml, Plastic bottle
CAS Number:  
Price
$118.85
Log in for availability
Each (500ml)
Product Details & Documents
Ammonia Ionic Strength Adjustor (ISA) Buffer Solution for ammonia determinations by electrode methods.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Specifications
Specifications
  • Type de conditionnement:
    Plastic bottle
  • Désignation:
    Ammonia Ionic Strength Adjustor (ISA)
  • Taille:
    500 ml