Replacement Tips for Three Star Tweezers, Straight Delrin Soft Replaceable Tip, Excelta | Avantor
Replacement Tips for Three Star Tweezers, Straight Delrin Soft Replaceable Tip, Excelta
Supplier:   Excelta
Replacement Tips for Three Star Tweezers, Straight Delrin Soft Replaceable Tip, Excelta
Supplier:  Excelta
Certificates
Replacement Tips for Three Star Tweezers, Straight Delrin Soft Replaceable Tip, Excelta
Replacement Tips for Three Star Tweezers, Straight Delrin Soft Replaceable Tip, Excelta
2 Options Available Below
Product Details & Documents
These soft replaceable tip tweezers feature Acetal tips which are electrically insulative and resistant to temperatures up to 185 °F.
The bodies are made of anti-magnetic Neverust® stainless steel (AISI 316L). Excellent for cleanroom applications and for avoiding the marring or scratching of items being handled.
All replacement tips come complete with 2 tips and 4 screws.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Tip Length
    Tip Style
    Tip Width
    Curvature
    Availability
    Price
  • Replacement tips for 159b-rtw, 3 star quality
    Acetal
    0.95 mm (0.038")
    Straight
    0.04" (1 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18.58
    $18.58
    Each
       $18.58
    Supplier Number: 159B-RTWX
  • Replacement tips for 159d-rtw, 3 star quality
    Acetal
    0.38 mm (0.015")
    Straight Tip
    0.06" (1.5 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18.34
    $18.34
    Each
       $18.34
    Supplier Number: 159D-RTWX