Tweezer Kitsets, Excelta Corp®
Supplier: Excelta
Certificates
About this item
Tweezer Kitsets contains of sets of stainless Steel, Anti-magnetic tweezers of various shapes and points.
- Ideal for engineers, variety of tip shapes
- Cleanroom safe, stainless steel
- Convenient wood case or pouch for storage
These tweezer kits are ideal for engineers. They offer a variety of different tweezer styles in a convenient pouch or wooden case. The tweezers are made of anti-magnetic stainless steel.
Frequently Bought Together