Falcon® 96-Multiwell Insert Systems, Corning | Avantor
Falcon® 96-Multiwell Insert Systems, Corning
Falcon™ 96-Multiwell Insert System with Feeder Tray and Lid, 96, 0.08 cm², 1.0 µm, Sterile, Tissue culture (TC)
Catalog # 10147-798
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  353938
Falcon® 96-Multiwell Insert Systems, Corning
Catalog # 10147-798
Supplier:  Corning
   Low Manufacturing Impact
Falcon® 96-Multiwell Insert Systems, Corning
Falcon® 96-Multiwell Insert Systems, Corning
Falcon™ 96-Multiwell Insert System with Feeder Tray and Lid, 96, 0.08 cm², 1.0 µm, Sterile, Tissue culture (TC)
Price
$2,270.00
Log in pour la disponibilité
Case of 5 Each
La quantité minimale de commande est de 1
Product Details & Documents
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1.0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
  • Automation compatible design suitable for use with most robots and fluid handling instruments
  • Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Ideal for drug transport studies
Spécifications
  • Nb de puits:
    96
  • Taille de pore:
    1.0 µm
  • Sterility:
    Sterile
  • Désignation:
    Falcon™ 96-Multiwell Insert System with Feeder Tray and Lid
  • Surface Treatment:
    Tissue culture (TC)
  • Cell Growth Area:
    0.08 cm²
  • Cat. No.:
    10147-798
  • Environmental certification:
    ISO 14001 - Bedford (reg. no. 14.4942)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Falcon™ 96 square well, angled bottom liquid receiver tray and lid
    Falcon™ 96 square well, angled bottom liquid receiver tray and lid
    BD-353925
    Log in pour la disponibilité
    Case of 5 - $192.00