PathoScreen Field Test Kit, Hach
PathoScreen Field Test Kit
Catalog # CA11014-402
Supplier: Hach
: 2859100
Jump to:
Product Details & Documents
PathoScreen Field Test Kit
Enough media for 100 presence-absence tests using 20 mL sample, or 20 MPN tests. Reusable glass vials, and foil-pillowed media, increases shelflife. No refrigeration required for media.
Specifications
Specifications
- Désignation:PathoScreen Field Test Kit