PathoScreen Field Test Kit, Hach
PathoScreen Field Test Kit
Catalog # CA11014-402
Supplier:   Hach
 :  2859100
PathoScreen Field Test Kit, Hach
Catalog # CA11014-402
Supplier:  Hach
CAS Number:  
PathoScreen Field Test Kit, Hach
PathoScreen Field Test Kit, Hach
PathoScreen Field Test Kit
CAS Number:  
Price
$252.99
Log in for availability
Each
Product Details & Documents
PathoScreen Field Test Kit
Enough media for 100 presence-absence tests using 20 mL sample, or 20 MPN tests. Reusable glass vials, and foil-pillowed media, increases shelflife. No refrigeration required for media.
Specifications
Specifications
  • Désignation:
    PathoScreen Field Test Kit