Sample Cells, Round Plastic, Hach | Avantor
Sample Cells, Round Plastic, Hach
Supplier:   Hach
Sample Cells, Round Plastic, Hach
Supplier:  Hach
Certificates
Sample Cells, Round Plastic, Hach
Sample Cells, Round Plastic, Hach
2 Options Available Below
Product Details & Documents
Round, plastic sample cells for use with Hach Spectrophotometers
Cells come in sets of two with caps.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Ø
    Pathlength
    Availability
    Price
  • Round Plastic Sample Cells, Set of 2
    5 and 10 ml
    1"
    1 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $102.32
    $102.32
    Each
       $102.32
    Supplier Number: 4165802
  • Round Plastic Sample Cells, Set of 2
    10 ml
    1"
    1 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $111.28
    $111.28
    Each
       $111.28
    Supplier Number: 4864302