Stablcal® Turbidity Standards Calibration Kit, 2100N/N IS Turbidimeter, 500 ml bottles, Hach
Catalog # CA97026-812
Supplier: Hach
CAS Number:
Specifications
- Turbidity:4000 NTU200 NTU< 0.1 NTU20 NTU1000 NTU
- Description:Stablcal® Turbidity Standards Calibration Kit
- Size:5×500 ml bottle
- Concentration:< 0.1, 20, 200, 1000, 4000 NTU
- Cat. no.:CA97026-812
- For Use With:2100N / N IS Turbidimeter
- Supplier No.:2662100
Specifications
About this item
Stablcal® Turbidity Standard for all Turbidimeters.
- Precise, Premixed Formazin Solutions
- Based on Formazin Primary Standards
- Enhanced Stability Saves Time and Labor
- Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of <0.1, 20, 200, 1000 and 4000 NTU standards.