LeadTrak™ Reagent Set, Fast Column Extraction Method, Hach
Reagent Set for Lead determination by the LeadTrak™ Fast Column Extraction
Hach Method 8317. Range: 5 to 150 µg/L as Pb. Sample size: 100 mL.
: 20 tests per reagent set. Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
- Méthode chimique:LeadTrak
- Désignation:LeadTrak™ Reagent Set, Fast Column Extraction Method, 20 tests
- Taille:100 mL
- Range:5 - 150 μg/l Pb
- No. of Tests:20
- Cat. No.:CA23750-00
- Instrument:All except DR/800
- Method:8317 - LeadTrak