Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Molybdate Test Kit, Model MO-2, Hach | Avantor

Molybdate Test Kit, Model MO-2, Hach

Molybdate Test Kit, Model MO-2, Mercaptoacetic acid, 0 - 40 mg/l Mo, 0.2 - 10.0 mg/l, 100
Catalog # CA11024-956
Supplier:   Hach
Fournisseur #:  1419301
Molybdate Test Kit, Model MO-2, Hach
Catalog # CA11024-956
Supplier:  Hach
CAS Number:  
Molybdate Test Kit, Model MO-2, Hach
Molybdate Test Kit, Model MO-2, Hach

Molybdate Test Kit, Model MO-2, Mercaptoacetic acid, 0 - 40 mg/l Mo, 0.2 - 10.0 mg/l, 100

CAS Number:  
Price
$977.45
Log in pour la disponibilité
Each
Product Details & Documents

Molybdates are gaining over chromates for use as corrosion inhibitors due to lower toxicity

The high cost of these treatments warrant careful control of their concentration. Kit reagents react with Molybdate to form a yellow color, which is measured with a color disc. Effectively monitors corrosion inhibitors.

Spécifications

Spécifications

  • Méthode chimique:
    Mercaptoacetic acid
  • Désignation:
    Molybdate Test Kit, Model MO-2
  • Range:
    0 - 40 mg/l Mo, 0.2 - 10.0 mg/l
  • No. of Tests:
    100
  • Cat. No.:
    CA11024-956
  • Model:
    MO-2
  • Case style:
    F
  • Increments:
    smallest steps - 1
  • Method:
    Color disc/ Mercaptoacetic acid

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.