About this item
The capacity of the Criterion™ Blotter is 2 midi-sized or 4 mini-sized gels
- Transfers in 30 min to 1 hr for most proteins
- Overnight transfer at lower voltages
- Assembly tray for gel soaking, sandwich assembly, and ensuring correct orientation of gels and blots
- Cooling unit is sufficient for absorbing heat during most transfers
- Color-coded, easy-lock cassettes
- Optional cooling coil for precise temperature control
- Overflow chambers keep the laboratory bench dry
Blotters are available with a choice of plate or wire electrodes. Durable plate electrodes consist of a platinum-coated titanium anode and a stainless steel cathode. Wire electrodes are platinum.
The close spacing of the electrodes (4.3 cm) provides a high field strength and allows rapid transfer with one or two cassettes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.