F422 HDPE Foam-Lined Polypropylene Caps, Qorpak®
Catalog # 89237-498
Supplier: Qorpak
Small Business Enterprise
Specifications
- Cap Size:38-439
- Cat. no.:89237-498
- Supplier No.:CAP-00379
Specifications
About this item
F422 HDPE Foam Lined Polypropylene Caps are acid resistant
This F422 liner is comprised of an 0.38" LDPE core sandwiched between identical layers of 0.002–0.003" virgin HDPE.