Paddle Tester, Total Aerobic Bacteria/Disinfection Control, Hach
CA97026-326
: Hach
:
- Description:Paddle Tester, Total Aerobic Bacteria/Disinfection Control
- Cat. no.:CA97026-326
- Supplier No.:2619510
Hach Paddle Testers allow fast, easy screening of water samples, solid surfaces, and nonviscous liquids where unsanitary conditions may exist.
- Save time and simplify lab and field testing with Hach prepared P/A media
- Report Total Coliform values with EPA accepted PresenceAbsence bottles and media
- Simplify routine testing with selfcontained, easytouse Paddle Testers and BART™ products
The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a molded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle Tester, Total Aerobic Bacteria/Disinfection Control (Tryptic Soy Agar/DE Neutralizing Agar).
: 10/pk.