Dial Barometer, Sper Scientific
Dial Barometer
Catalog # 61220-786
Supplier: Sper Scientific
Fournisseur #: 736930
Jump to:
Product Details & Documents
Dial barometer for classroom, lab and industrial use.
Large, clear dual-scale barometer in hPa (hecto Pascal) and mm (millimeter mercury) on a 4" dial which is easy to read from a distance. For classroom, lab, or industrial use. Wall mounts.
Ordering information: 5 year warranty.
Specifications
- Désignation:Dial Barometer
- Cat. No.:61220-786
- Accuracy:±3 to 4 hPa
- Weight:2.6 oz.