Borate Buffer Solution, 1 L, Hach
CA97026-634
: Hach
:
- Description:Borate Buffer Solution
- Size:1 L
- Cat. no.:CA97026-634
- Supplier No.:1470953
For Nitrogen-Ammonia distillation
APHA formulation. 1 L.
For Nitrogen-Ammonia distillation
APHA formulation. 1 L.