Spécifications
- Frequency:43,000 Hz
- Max. output power:35 W
- Capacity:600 ml (0.5 qt.) tank
- Description:Mini Ultrasonic Cleaner
- Electrical:100/110V AC
- W×D×H:203×117×127 mm (8 × 4.5 × 5")
- Cycle time:3 min
- Tank W×D×H:165×89×64 mm (6.5 × 3.5 × 2.5")
- Weight:2 lbs.
- Cat. no.:14003-062
- Supplier No.:100005
Spécifications
À propos de cet article
Compact Ultrasonic Cleaner for Jewelry, Eyeglasses, Instruments and More.
The Mini Ultrasonic Cleaner is designed to remove grease, dust or other contaminants from surfaces and cavities of small objects, parts and samples such as cuvettes, lenses, glassware, jewelry, stainless steel instruments, metals and more. Simply place objects directly into the stainless steel tank and press “on”. The unit generates thousands of tiny cleansing bubbles which vibrate at a very high electrical frequency. When the vibration speed rises above the ultrasonic frequency level, bubbles explode and generate strong power, cleaning the surfaces and cavities of hard-to-clean objects. Your samples will be clean in 3 minutes after which the unit will shut off automatically.
Ultrasonic cleaning does not scratch the surface or harm objects like many chemical or abrasive cleansers. Not intended for commercial, non-stop use. Includes a plastic basket and bracelet holder.