Falcon® Snap Caps for Test Tubes, Corning
Catalog # CA60819-251
Supplier: Corning
Specifications
- Sterility:Sterile
- Description:Falcon™ Snap Caps
- Size:12 mm
- Cat. no.:CA60819-251
- Packaging:500/Pack
- Fits:12×75 mm Test Tube
- Supplier No.:352032
Specifications
About this item
Falcon® snap caps are designed for use with 12×75 mm test tubes
These polyethylene caps feature a patented, two-position fit, loose for aerobic culturing, tight for sealing. Sterile.
Ordering information: Snap caps are packed 500 per pack, 2000 per case.