Torque Meter, Sper Scientific
Torque Meter, Sper Scientific
Supplier:   Sper Scientific
Torque Meter, Sper Scientific
Supplier:  Sper Scientific
Certificates
Torque Meter, Sper Scientific
Torque Meter, Sper Scientific
2 Options Available Below
Product Details & Documents
This portable meter is used for bolt torqueing, torque wrench calibration, switch or contact closure measurement and other industrial, QC, and materials testing applications
Features 3 units of measure as well as hold, peak, min/max, fast/slow sampling, push button hi/low resolution selector, auto power off, and RS232 computer interface. Comes in a hard carrying case with a 15 kg/cm torque probe with 4.5’ (140mm) cable and a 9V battery.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-944.
Ordering information: 1 yr. warranty.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Torque Meter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,771.55
    $1,771.55
    Each
       $1,771.55
    Supplier Number: 840062
  • Torque Meter Certified
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,310.41
    $2,310.41
    Each
       $2,310.41
    Supplier Number: 840062C