Continuous Alarm Timer, Sper Scientific | Avantor
Continuous Alarm Timer, Sper Scientific
Supplier:   Sper Scientific
Continuous Alarm Timer, Sper Scientific
Supplier:  Sper Scientific
Certificates
Continuous Alarm Timer, Sper Scientific
Continuous Alarm Timer, Sper Scientific
2 Options Available Below
Product Details & Documents
Select either a bright flashing yellow LED visual alarm or an audible alarm.
A memory recalls the last timer setting for reuse. Built in stand for bench top use, clips to a pocket, or sticks to any metal surface with a magnetic back.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-906.
Ordering information: Five year warranty.
Delivery information: Two AAA batteries included.
Product Family Options
Informations sur le produit
  • Désignation
    Minuterie
    W×H×D
    Weight
    Availability
    Price
  • Continuous alarm timer
    99 min
    75×55×15 mm (3×2×⅝")
    57 g (2 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $89.72
    $89.72
    Each
       $89.72
    Supplier Number: 810045
  • Continuous alarm timer certified
    75×55×15 mm (3×2×⅝")
    57 g (2 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $223.73
    $223.73
    Each
       $223.73
    Supplier Number: 810045C