Visual Alarm Timer 99 min, Sper Scientific
Compact visual and audible alarm timer.
- A memory recalls the last timer setting for reuse
- Built in stand for bench top use
Select either a bright flashing yellow LED visual alarm or an audible alarm. Often used by testing organizations, including Law School Admissions Council (LSAC). Features clips to a pocket, or sticks to any metal surface with a magnetic back.
: N.I.S.T. traceable certificate of compliance for 97035-890.
: Five year warranty.
: Two AAA batteries included.
Frequently Bought Together