Leakbuster™ 3 Specimen Container, Starplex®
Fournisseur: Starplex
Certificats
À propos de cet article
This new specimen container features a tamper evident cap that ensures proper cap application by providing visual alignment points, as well as audible and distinct tactile closure confirmation
The textured, flat cap provides an additional writing surface in a stackable format. Each container is lot numbered and features clear graduations.
Ordering information: Leakbuster™ 3 is also available in a clear plastic O.R. wrap format and a Midstream Kit format with a castille wipe enclosed in a clear plastic peel pack.
Frequently Bought Together