IntelliCAL™ MTC101 Standard Gel Filled ORP Electrodes, Hach
Fournisseur: Hach
Certificats
À propos de cet article
IntelliCAL™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor.
- Provides fast, stable response in a variety of applications
- Ultimate traceability in measurement history
- User alerted when recalibration is needed
- Flat disc sensor for easy cleaning
- Low maintenance
- Ag/AgCl reference system
- Can be moved between meters without the need to recalibrate or reenter measurement settings
The standard version is available with a 1 or 3 meter cable and is intended for laboratory use. The MTC101 is ideal for measuring absolute mV values in wastewater, drinking water and general applications.
Weight: 0.17 lbs. (0.076 kg)
Frequently Bought Together