Sodium Thiosulfate Standard Solution, Stabilized, 0.0246 N, 100 ml, Hach
Catalogue # CA24092-32
Fournisseur: Hach
Numéro CAS:
Spécifications
- Description:Sodium Thiosulfate Standard Solution, Stabilized, 0.0246 N
- Size:100 ml Marked dropping bottle
- Method:Iodometric Titration
- Cat. no.:CA24092-32
- Supplier No.:24092-32
- No. of Tests:Varies
Spécifications
À propos de cet article
Stabilized sodium thiosulfate titrant for chlorine determination by iodometric titration.