Potassium hydroxyde solution standard 8.00 N | Avantor
Potassium hydroxyde solution standard 8.00 N
Dropping bottle, marked, 100 ml
Catalog # CA282-32H
Corrosif
Irritant
Supplier:   Hach
Fournisseur #:  28232H
Potassium hydroxyde solution standard 8.00 N
Catalog # CA282-32H
Supplier:  Hach
CAS Number:  1310-58-3
Potassium hydroxyde solution standard 8.00 N
Potassium hydroxyde solution standard 8.00 N
Dropping bottle, marked, 100 ml
CAS Number:  1310-58-3
Price
$43.34
Log in pour la disponibilité
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For pH adjustment in calcium determination by EDTA titration
  • MDB botle
Hach methods 8222 (buret) and 8204 (Digital Titrator).
Synonymes:  Potasse caustique
  • CAS Number:1310-58-3
  • Formule:KOH
  • Numéro MDL:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Température de conservation:Température ambiante
  • ONU:1814
  • MARC:8,II
Spécifications
Spécifications
  • Type de conditionnement:
    Dropping bottle, marked
  • Taille:
    100 ml