Flask, Cylindrical, Mini-Lab, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
This round bottom, cylindrical flask is useful in many Mini-Lab assemblies, and in other applications where a wide-mouth flask of small capacity is desired
For 89074-568 use mantle 89075-024; for 89074-570 and 89074-574 use mantle 89075-026; and for 89074-572 use mantle 89075-028.
Use with 89074-562, 89074-564, or 89074-566 heads. Fabricated from borosilicate glass.
Ordering information: 100 mL flask (only) is available with ground or polished joint.
Frequently Bought Together