PFA Jars
À propos de cet article
Used for the collection, storage, or disposition of a wide variety of solutions or solids
Service temperature range is –200 to +260 °C (–328 to +500 °F).
The seamless design, radiused interior corners, and wide mouth opening allow for easy transfer and removal of contents. Knurled closures enable a secure grip. A proprietary PFA molding process produces a container with strong yet translucent walls so contents are visible.
Ordering information: Closures are not included, and must be ordered separately.
7 Options Available Below
Product Details & Documents
Used for the collection, storage, or disposition of a wide variety of solutions or solids
Service temperature range is –200 to +260 °C (–328 to +500 °F).
The seamless design, radiused interior corners, and wide mouth opening allow for easy transfer and removal of contents. Knurled closures enable a secure grip. A proprietary PFA molding process produces a container with strong yet translucent walls so contents are visible.
Ordering information: Closures are not included, and must be ordered separately.
Product Family Options
Informations sur le produit
- DésignationCapacitéHauteurØAvailabilityPrice
- Supplier Number: 100-0090-01
- Supplier Number: 100-0240-01
- Supplier Number: 100-1000-01
- Supplier Number: 100-0120-01Spécifications:New ProductNew Product
- Supplier Number: 100-0180-01Spécifications:New ProductNew Product
- Supplier Number: 100-0180-02Spécifications:New ProductNew Product
- Supplier Number: 100-0350-01Spécifications:New ProductNew Product
Accessories
Informations sur le produit
- Catalog #HeightØAvailabilityPrice
- PFA Closure, 53mm, for use with 30617-474PFA Closure, 53mm, for use with 30617-47430617-51619.1 mm (0.8")62.5 mm (2.5")Log in pour la disponibilitéEach - $122.10
Informations sur le produit
- Catalog #HeightØAvailabilityPrice
- PFA Closure, 70mm, for use with 30617-476PFA Closure, 70mm, for use with 30617-47630617-51819.6 mm (0.8")80 mm (3.1")Log in pour la disponibilitéEach - $147.32
Informations sur le produit
- Catalog #HeightØAvailabilityPrice
- PFA Closure, 110mm, for use with 30617-480PFA Closure, 110mm, for use with 30617-48030617-52229 mm (1.1")122.7 mm (4.8")Log in pour la disponibilitéEach - $497.77
Informations sur le produit
- Catalog #HeightØAvailabilityPrice
- PFA Closure, 89mm, for use with 30617-478PFA Closure, 89mm, for use with 30617-47830617-52023.1 mm (0.9")99.8 mm (3.9")Log in pour la disponibilitéEach - $237.09