Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
Phosphorus (Reactive) TNT Reagent Set, Low Range, Ascorbic acid, 0.06 - 5.00 mg/l PO₄, 25 - 50
Supplier: Hach
Fournisseur #: 2742545
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
Catalog # CA97026-534
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
For determination of Reactive Phosphorus (ortho-phosphate) by the PhosVer® 3 Ascorbic Acid Test 'N Tube™ method.
- Test 'N Tube products provide safe, convenient testing
- Easy ordering
Hach Method 8048. Range: 0.06 - 5.00 mg/L PO4. Set includes: PhosVer® 3 Phosphate Reagent Powder Pillows and Reactive Phosphorus Test 'N Tube Dilution Vials for up to 50 tests.
Spécifications
Spécifications
- Méthode chimique:Ascorbic acid
- Désignation:Phosphorus (Reactive) TNT Reagent Set, Low Range
- Range:0.06 - 5.00 mg/l PO₄
- No. of Tests:25 - 50
- Cat. No.:CA97026-534
- Instrument:All except DR/820, PC II
- Method:8048 - Ascorbic Acid