À propos de cet article
Can cancer be inherited?
- NGSS Life Science DCIS LS1 and LS3
- Assessments and vocabulary words target Common Core standard
- Pre and post-laboratory assessments
- Links to real world concepts
- Estimated Lab Time: 80 minutes over one to two days
- Aligned to NGSS standards (MS/HS: LS1, LS3)
Nearly a quarter of a million women in the US will develop breast cancer in a given year. Cancers may be caused by environmentally-induced or inherited genetic mutations. In this activity students will perform agarose gel electrophoresis as a molecular diagnostic test for hereditary breast cancer. Students will use the test results to predict the patient’s risk of developing cancer. Students will also learn how to analyze a family pedigree all while employing NGSS Life Science Disciplinary Core Ideas LS1 and LS3 (From Molecules to Organisms: Structures and Processes and Heredity: Inheritance and Variations of Traits). This kit includes enough materials to run 3 gels.
Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with scientific standards explored. A student copymaster is also included.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.