WorkWipes® Reclaimed Colored Sweatshirt in Bag, New Pig
Catalog # 89073-924
Supplier: New Pig
Specifications
- Cat. no.:89073-924
- Packaging:Compressed
- Weight:11.3 kg (25 lbs.)
- Supplier No.:WIP569
Specifications
About this item
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Thick, absorbent rags are ideal for heavy-duty wiping
- An excellent combination of softness and strength
Eco- and cost-friendly rags are made from recycled, post-consumer cotton and cotton blend sweatshirt material.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Ordering information: Scanned for metal. Not sorted for printing. Not washed.