WorkWipes® Reclaimed White Healthcare Linen in Bag, New Pig
Catalog # 89239-110
Supplier: New Pig
Specifications
- Cat. no.:89239-110
- Packaging:Compressed
- Weight:25 lbs.
- Supplier No.:WIP594
Specifications
About this item
Strict material and size requirements mean you get more usable, high quality rags in every package.
- White color is ideal for use with solvents, which might cause colored rags to bleed
Eco- and cost-friendly rags are made from white, post-consumer recycled cotton and cotton blend healthcare sheets and pillow cases. Low-lint, laundered material is ideal for staining, wiping, polishing and other general tasks.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Ordering information: Scanned for metal. Not sorted for printing. Washed.