White Large Display Timer, Sper Scientific | Avantor
White Large Display Timer, Sper Scientific
Supplier:   Sper Scientific
White Large Display Timer, Sper Scientific
Supplier:  Sper Scientific
Certificates
White Large Display Timer, Sper Scientific
White Large Display Timer, Sper Scientific
2 Options Available Below
Product Details & Documents
This easy-to-read, large display timer features the largest display available on a timer: 3×2¹/₂"
  • Can be easily read from across a room
  • White case has extra large start/stop button and a loud alarm
  • Wall mount or fold for bench top use
  • Runs for approximately two years on one AAA battery
Simultaneously displays either one count up channel and a clock, or two count down channels. All channels have a range of 24 hours with 1 second resolution. Also indicates how much time has elapsed after a count down alarm sounds and conveniently recalls the last timer setting for reuse.
Certifications: N.I.S.T. traceable certificate of compliance for certified option.
Ordering information: One AAA battery included. Five Year Warranty.
Product Family Options
Informations sur le produit
  • Désignation
    W×H×D
    Weight
    Availability
    Price
  • White large display timer
    5×3¾×½"
    6.7 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $153.19
    $153.19
    Each
       $153.19
    Supplier Number: 810005
  • White large display timer, certified
    5×3¾×½"
    6.7 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $284.98
    $284.98
    Each
       $284.98
    Supplier Number: 810005C